Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3',4',5-Trichlorosalicylanilide 98.0+%, TCI America™
SDP

Catalog No. T066810G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T066810G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T066810G Supplier TCI America Supplier No. T066810G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 642-84-2
Molecular Formula C13H8Cl3NO2
Molecular Weight (g/mol) 316.56
MDL Number MFCD00059622
InChI Key RSJBLPJKXGNMFW-UHFFFAOYSA-N
PubChem CID 69506
IUPAC Name 5-chloro-N-(3,4-dichlorophenyl)-2-hydroxybenzamide
SMILES OC1=CC=C(Cl)C=C1C(=O)NC1=CC=C(Cl)C(Cl)=C1

Specifications

Color White-Yellow
Quantity 10 g
Formula Weight 316.56
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3′,4′,5-Trichlorosalicylanilide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.