Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3,4,5-Trifluorobenzaldehyde

Catalog No. 5291765ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
52-917-65ML 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 52-917-65ML Supplier Sigma Aldrich Supplier No. 5291765ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

3,4,5-Trifluorobenzaldehyde

Specifications

CAS 132123-54-7
Density 1.42 g/mL (at 25°C (literature))
Boiling Point 174°C (lit.)
Molecular Formula C7H3F3O
Refractive Index n20/D 1.482 (literature)
Linear Formula F3C6H2CHO
MDL Number MFCD00083523
Quantity 1 g
Molecular Weight (g/mol) 160.09
Percent Purity 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.