Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4,5-Trifluorobenzoic Acid 98.0+%, TCI America™
SDP

Catalog No. T16405G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T16405G 5 g
T16401G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. T16405G Supplier TCI America Supplier No. T16405G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 121602-93-5
Molecular Formula C7H3F3O2
Molecular Weight (g/mol) 176.094
MDL Number MFCD00074962
InChI Key VJMYKESYFHYUEQ-UHFFFAOYSA-N
Synonym 3,4,5-trifluorobenzoicacid, benzoic acid, 3,4,5-trifluoro, 3,4,5-trifluoro benzoic acid, 3,4,5-trifluoro-benzoic acid, acmc-209aen, ksc177i9t, rarechem al bo 0682, 3,4,5-trifluorobenzoyl alcohol, buttpark 30\01-46
PubChem CID 853154
IUPAC Name 3,4,5-trifluorobenzoic acid
SMILES C1=C(C=C(C(=C1F)F)F)C(=O)O

Specifications

Melting Point 98°C
Color White
Quantity 5 g
Formula Weight 176.09
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3,4,5-Trifluorobenzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.