Learn More
Sigma Aldrich 3,4,5-trifluorophenylacetylene
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
- Anisaldehyde dimethyl acetal is a clear, colorless to light yellow liquid
- The flash point for anisaldehyde dimethyl acetal is 114.0c
- It undergoes allylation reactions with allyltrimethylsilane catalysed by iron(iii) chloride
- Anisaldehyde dimethyl acetal undergoes ytterbium-catalyzed tandem carboalkoxylation/friedel-crafts reaction with 1-phenylbenzylidenecyclopropane to yield 1-(p-anisyl)-2-(2-methoxyethyl)-3-phenylindene.anisaldehyde dimethyl acetal was used in the synthesis of lipid a mimic precursor
- It was also used in the synthesis of 2,5-dioxopyrrolidin-1-yl-3-((4r)-2-(4-methoxyphenyl)-5,5-dimethyl-1,3-dioxane-4-carboxamido)propanoate.protecting group reagent for diols, used e.g
- In carbohydrate chemistry; the acetal protection can be selectively cleaved under mild conditions
Specifications
Specifications
| Molecular Formula | C8H3F3 |
| Linear Formula | C8H3F3 |
| MDL Number | MFCD08458177 |
| Quantity | 50 mL |
| Molecular Weight (g/mol) | 156.1 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.