Learn More
Medchemexpress LLC 3,4,5-trimethoxy-N-{2-[3-(piperazin-1-ylmethyl)imidazo[2,1-b]thiazol-6-yl]phenyl}benzamide | 925432-73-1 | MFCD26406722 | 98.2% | 507.60 g·mol-1 | C26H29N5O4S | 100MG
SDP
Catalog No. 5000237785
Encompass_Preferred
SRT 1460 is a small-molecule activator of SIRT1 provided for research use in biochemical and cell-based studies. It activates SIRT1 with an EC1.5 of 2.9 μM, shows strong selectivity versus SIRT2 and SIRT3 (EC1.5 > 300 μM), and is supplied as an off-white to light yellow solid with solubility ≈10 mg/mL in DMSO.
  • Potent SIRT1 activation (EC1.5 = 2.9 μM).
  • High selectivity versus SIRT2 and SIRT3 (EC1.5 > 300 μM).
  • Off-white to light yellow solid appearance.
  • Soluble in DMSO for convenient stock preparation (≈10 mg/mL).
  • Stable when stored frozen as powder for extended periods.
  • Suitable for biochemical and cell-based assays.
Catalog No. 50-002-37785 Supplier Medchemexpress LLC Supplier No. HY124037100MG
SRT 1460 is a small-molecule activator of SIRT1 provided for research use in biochemical and cell-based studies. It activates SIRT1 with an EC1.5 of 2.9 μM, shows strong selectivity versus SIRT2 and SIRT3 (EC1.5 > 300 μM), and is supplied as an off-white to light yellow solid with solubility ≈10 mg/mL in DMSO.
  • Potent SIRT1 activation (EC1.5 = 2.9 μM).
  • High selectivity versus SIRT2 and SIRT3 (EC1.5 > 300 μM).
  • Off-white to light yellow solid appearance.
  • Soluble in DMSO for convenient stock preparation (≈10 mg/mL).
  • Stable when stored frozen as powder for extended periods.
  • Suitable for biochemical and cell-based assays.