Learn More
Medchemexpress LLC 3,4,5-trimethoxy-N-(2-methoxyethyl)-N-(4-phenyl-2-thiazolyl)benzamide | 461000-66-8 | MFCD03115431 | 98.9% | 428.50 | C22H24N2O5S | 25 MG
SDP
Supplier:  Medchemexpress LLC HY10336825MG
Encompass_Preferred
Eact is a small-molecule research reagent that selectively activates the calcium-activated chloride channel TMEM16A (ANO1). It also directly activates TRPV1 channels in sensory nociceptors, producing itch, acute nociception, and thermal hypersensitivity; supplied for in vitro research use.
  • Selective activator of TMEM16A (ANO1).
  • Direct activator of TRPV1 channels in sensory nociceptors.
  • High purity (≈98.9%) white to off-white solid for research applications.
  • Soluble in DMSO (50 mg/mL); ultrasonic assistance recommended.
  • Store powder at -20°C for long-term stability; in solvent store at -80°C.
Catalog No. 50-002-44702