Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3',4',5'-Trimethoxyacetophenone

Catalog No. T6810125G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T6810125G 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T6810125G Supplier Sigma Aldrich Supplier No. T6810125G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Triethylene glycol monobutyl ether (tegmbe) can be used as a solvent:
  • In the synthesis of superparamagnetic iron oxide nanoparticles by thermal decomposition method
  • In the removal of co2 due to its solubilize co2.it can also be used as a reactant in the preparation of polyether adducts of bis(1,1,1,5,5,5-hexafluoro-2,4-pentanedionato)barium

Specifications

CAS 1136-86-3
Melting Point 78°C to 80°C (lit.)
Boiling Point 173°C to 174°C (10 mmHg)
Molecular Formula C11H14O4
Linear Formula (CH3O)3C6H2COCH3
MDL Number MFCD00008738
Quantity 1 L
Molecular Weight (g/mol) 210.23
Percent Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.