Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3,4,5-Trimethoxybenzoyl chloride

Catalog No. T6980925G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T6980925G 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T6980925G Supplier Sigma Aldrich Supplier No. T6980925G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • This product is informer compound x1 of the aryl halide chemistry informer library developed by chemists at merck & co., inc., kenilworth, nj, u.s., which contains 18 drug-like molecules representative of those encountered in complex synthesis
  • By screening a new reaction against the informer library, chemists can directly compare and analyse a reaction's successes and shortcomings among different methods and various research teams
  • It may also be used to facilitate deeper method development for performance or utility.chemistry informer libraries: a chemoinformatics enabled approach to evaluate and advance synthetic methods, chem
  • Sci., 2016, 7, 2604-2613

Specifications

CAS 4521-61-3
Melting Point 81°C to 84°C (lit.)
Boiling Point 185°C (18 mmHg)
Molecular Formula C10H11ClO4
Linear Formula (CH3O)3C6H2COCl
MDL Number MFCD00000675
Quantity 100 mg
Molecular Weight (g/mol) 230.64
Percent Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.