Learn More
Medchemexpress LLC 3,4,5-Trimethoxycinnamic acid | 90-50-6 | 99.9% | 238.24 | 25 G
SDP
Catalog No. 5000366801
Encompass_Preferred
3,4,5-Trimethoxycinnamic acid is a phenylpropanoid isolated from the roots of *Polygala tenuifolia WILLD*. It has an anti-stress effect and can prolong sleeping time in animals. This compound increases the expression of GAD65 and the γ-subunit of the GABAA receptor, but it does not affect the amounts of α- and β-subunits.
  • Anti-stress effect
  • Prolongs sleeping time in animals
  • Increases expression of GAD65 and γ-subunit of GABAA receptor
  • Exhibits sedative effect in rats
  • Significantly decreases locomotor activity in mice
  • Available in solid form, white to off-white color
Catalog No. 50-003-66801 Supplier Medchemexpress LLC Supplier No. 25GHYW012123
3,4,5-Trimethoxycinnamic acid is a phenylpropanoid isolated from the roots of *Polygala tenuifolia WILLD*. It has an anti-stress effect and can prolong sleeping time in animals. This compound increases the expression of GAD65 and the γ-subunit of the GABAA receptor, but it does not affect the amounts of α- and β-subunits.
  • Anti-stress effect
  • Prolongs sleeping time in animals
  • Increases expression of GAD65 and γ-subunit of GABAA receptor
  • Exhibits sedative effect in rats
  • Significantly decreases locomotor activity in mice
  • Available in solid form, white to off-white color