Learn More
Medchemexpress LLC 3,4,5-Trimethoxycinnamic acid | 90-50-6 | 99.96% | 238.24 | 1 ML
SDP
Catalog No. 5000452329
Encompass_Preferred
3,4,5-Trimethoxycinnamic acid is a phenylpropanoid isolated from the roots of *Polygala tenuifolia WILLD*. It exhibits an anti-stress effect and prolongs sleeping time in animals. This compound increases the expression of GAD65 and the γ-subunit of the GABAA receptor, but does not affect the amounts of α- or β-subunits. It is for research use only.
  • Anti-stress effect
  • Prolongs sleeping time in animals
  • Increases expression of GAD65 and γ-subunit of GABAA receptor
  • For research use only
Catalog No. 50-004-52329 Supplier Medchemexpress LLC Supplier No. HYW01212310MM/1ML
3,4,5-Trimethoxycinnamic acid is a phenylpropanoid isolated from the roots of *Polygala tenuifolia WILLD*. It exhibits an anti-stress effect and prolongs sleeping time in animals. This compound increases the expression of GAD65 and the γ-subunit of the GABAA receptor, but does not affect the amounts of α- or β-subunits. It is for research use only.
  • Anti-stress effect
  • Prolongs sleeping time in animals
  • Increases expression of GAD65 and γ-subunit of GABAA receptor
  • For research use only