Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4,5-Trimethoxyphenylacetonitrile, 97%, Thermo Scientific™

Catalog No. AC151061000
Change view
Click to view available options
100g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC151061000 100g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC151061000 Supplier Thermo Scientific Chemicals Supplier No. 151061000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 13338-63-1
CAS Min % 96.0
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C11H13NO3
Linear Formula (CH3O)3C6H2CH2CN
Quantity 100g
MDL Number MFCD00001912
Synonym 3,4,5-trimethoxyphenylacetonitrile, 2-3,4,5-trimethoxyphenyl acetonitrile, 3,4,5-trimethoxybenzyl cyanide, 3,4,5-trimethoxybenzylnitrile, 3,4,5-trimethoxybenzeneacetonitrile, benzeneacetonitrile, 3,4,5-trimethoxy, 3,4,5-trimethoxyphenylacetylnitryl, 3,4,5-trimethoxyphenyl acetonitrile, acetonitrile, 3,4,5-trimethoxyphenyl, 2-3,4,5-trimethoxyphenyl ethanenitrile
InChI Key ACFJNTXCEQCDBX-UHFFFAOYSA-N
SMILES COC1=CC(CC#N)=CC(OC)=C1OC
IUPAC Name 2-(3,4,5-trimethoxyphenyl)acetonitrile
Molecular Weight (g/mol) 207.23
PubChem CID 25887
Formula Weight 207.23
Percent Purity ≥96.0%
Chemical Name or Material 3, 4, 5-Trimethoxyphenylacetonitrile, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Harmful in contact with skin.
Harmful if inhaled.
May cause respiratory irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 236-388-5
RTECSNumber AM2475000
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.