Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

[[(3,4,5-Trimethyl-1H-pyrrol-2-yl)(3,4,5-trimethyl-2H-pyrrol-2-ylidene)methyl]carbonitrile](difluoroborane) 98.0+%, TCI America™
SDP

Catalog No. T3127200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T3127200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T3127200MG Supplier TCI America Supplier No. T3127200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 157410-23-6
Molecular Formula C16H18BF2N3
Molecular Weight (g/mol) 301.148
InChI Key LXNVCQRKLGCOPW-UHFFFAOYSA-N
Synonym Pyrromethene 650
PubChem CID 20724664
SMILES [B-]1(N2C(=C(C(=C2C(=C3[N+]1=C(C(=C3C)C)C)C#N)C)C)C)(F)F

Specifications

Melting Point 255°C
Quantity 200 mg
Formula Weight 301.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material [[(3,4,5-Trimethyl-1H-pyrrol-2-yl)(3,4,5-trimethyl-2H-pyrrol-2-ylidene)methyl]carbonitrile](difluoro
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.