Learn More
Cayman Chemical 3,4,5-Trimethoxybenzoic Acid, Purity: ≥98%, CAS: 118-41-2, 250 g
SDP
Catalog No. 502910245
Encompass_Preferred
An active metabolite of trimebutine; active against S. aureus (MIC = 0.97 µg/ml)
Catalog No. 50-291-0245 Supplier Cayman Chemical Supplier No. 34750250 G
An active metabolite of trimebutine; active against S. aureus (MIC = 0.97 µg/ml)