Learn More
Cayman Chemical 3 4 5Trismethoxybenzophenone
SDP
Catalog No. 502889288
Encompass_Preferred
An analog of trimethoxy resveratrol; inhibits proliferation of a variety of human tumor cell lines at 0.4-2 UG/ml
Catalog No. 50-288-9288 Supplier Cayman Chemical Supplier No. 1000418510 MG
Add to Cart
Add to Cart
An analog of trimethoxy resveratrol; inhibits proliferation of a variety of human tumor cell lines at 0.4-2 UG/ml