Learn More
Medchemexpress LLC 3-[4-(6-(3-(dimethylamino)propoxy)benzo[d]oxazol-2-yl)phenoxy]-N,N-dimethylpropan-1-amine | 1219962-49-8 | >98.0% | 397.52 g/mol | C23H31N3O3 | 250MG
SDP
Catalog No. 5000236450
Encompass_Preferred
AT791 is a small-molecule inhibitor of Toll-like receptors TLR7 and TLR9 used in research to modulate innate immune signaling. It inhibits TLR7 and TLR9 signaling in human and mouse cell types, prevents DNA-TLR9 interaction in vitro, and has demonstrated activity in mouse challenge models. Supplied as a research-grade powder; verify lot documentation for purity and handling.
  • Potent inhibitor of TLR7 and TLR9.
  • Demonstrated activity in human and mouse cell types.
  • Blocks DNA-TLR9 interaction in vitro.
  • Shows efficacy in mouse in vivo challenge models.
  • Research-grade powder with >98.0% purity by HPLC.
  • Chemical formula C23H31N3O3 and molecular weight ≈397.52 g/mol.
Catalog No. 50-002-36450 Supplier Medchemexpress LLC Supplier No. HY124603250MG
AT791 is a small-molecule inhibitor of Toll-like receptors TLR7 and TLR9 used in research to modulate innate immune signaling. It inhibits TLR7 and TLR9 signaling in human and mouse cell types, prevents DNA-TLR9 interaction in vitro, and has demonstrated activity in mouse challenge models. Supplied as a research-grade powder; verify lot documentation for purity and handling.
  • Potent inhibitor of TLR7 and TLR9.
  • Demonstrated activity in human and mouse cell types.
  • Blocks DNA-TLR9 interaction in vitro.
  • Shows efficacy in mouse in vivo challenge models.
  • Research-grade powder with >98.0% purity by HPLC.
  • Chemical formula C23H31N3O3 and molecular weight ≈397.52 g/mol.