Learn More
Medchemexpress LLC 3,4,6-tri-O-benzyl-D-galactal | 80040-79-5 | 416.51 g/mol | C27H28O4 | 1 G
SDP
Catalog No. 5000208758
Encompass_Preferred
3,4,6-tri-O-benzyl-D-galactal is a benzyl-protected derivative of D-galactal supplied as a powder for use in glycobiology and carbohydrate-chemistry research.
  • protected at the 3, 4, and 6 positions with benzyl groups
  • used as a building block in carbohydrate synthesis and biochemical assays
  • supplied as a powder suitable for laboratory handling
  • molecular weight 416.51 g/mol; formula C27H28O4
Catalog No. 50-002-08758 Supplier Medchemexpress LLC Supplier No. HYW0901031G
3,4,6-tri-O-benzyl-D-galactal is a benzyl-protected derivative of D-galactal supplied as a powder for use in glycobiology and carbohydrate-chemistry research.
  • protected at the 3, 4, and 6 positions with benzyl groups
  • used as a building block in carbohydrate synthesis and biochemical assays
  • supplied as a powder suitable for laboratory handling
  • molecular weight 416.51 g/mol; formula C27H28O4