Learn More
Medchemexpress LLC 3,4,6-tri-O-benzyl-D-galactal | 80040-79-5 | MFCD09039276 | 98.9% | 416.51 g/mol | C27H28O4 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW090103
Encompass_Preferred
3,4,6-Tri-O-benzyl-D-galactal is a benzyl-protected derivative of D-galactal, used as a specialized intermediate and research reagent in glycobiology and synthetic carbohydrate chemistry.
  • common applications include carbohydrate synthesis and glycobiology research.
  • listed with CAS number 80040-79-5, formula C27H28O4, and molecular weight 416.51 g/mol.
  • available as a powder and supplied in 25 g quantities.
  • analytical documentation such as datasheet, COA, and spectral data (HNMR, RP-HPLC) are available.
  • store as a powder under recommended refrigerated conditions to maintain stability.
Catalog No. 50-002-08840