Learn More
Medchemexpress LLC 3,4,7,8-Tetramethyl-1,10-phenanthroline | 1660-93-1 | 99.8% | 236.31 | 10 G
SDP
Catalog No. 5000404337
Encompass_Preferred
3,4,7,8-Tetramethyl-1,10-phenanthroline (TMPhen) is an organic molecule that functions as a ligand or catalyst. It is utilized across various fields, including organometallic chemical reactions, electrochemical detection, and organic optoelectronic devices. Its significant performance in fluorescent probes, biosensors, and photocatalytic reactions has led to its extensive use in chemistry and life sciences research.
  • Used as a ligand or catalyst
  • Applications in organometallic chemical reactions
  • Used for electrochemical detection
  • Applicable in organic optoelectronic devices
  • Significant performance in fluorescent probes
  • Used in biosensors
  • Suitable for photocatalytic reactions
  • Intended for research use only
Catalog No. 50-004-04337 Supplier Medchemexpress LLC Supplier No. HY3451510G
3,4,7,8-Tetramethyl-1,10-phenanthroline (TMPhen) is an organic molecule that functions as a ligand or catalyst. It is utilized across various fields, including organometallic chemical reactions, electrochemical detection, and organic optoelectronic devices. Its significant performance in fluorescent probes, biosensors, and photocatalytic reactions has led to its extensive use in chemistry and life sciences research.
  • Used as a ligand or catalyst
  • Applications in organometallic chemical reactions
  • Used for electrochemical detection
  • Applicable in organic optoelectronic devices
  • Significant performance in fluorescent probes
  • Used in biosensors
  • Suitable for photocatalytic reactions
  • Intended for research use only