Learn More
Medchemexpress LLC 3,4,7,8-Tetramethyl-1,10-phenanthroline | 1660-93-1 | 99.8% | 236.31 | 100 G
SDP
Catalog No. 5000404235
Encompass_Preferred
3,4,7,8-Tetramethyl-1,10-phenanthroline (TMPhen) is an organic molecule commonly used as a ligand or catalyst. It has a wide range of applications in different fields and is utilized in research in chemistry and life sciences.
  • Applicable in organometallic chemical reactions.
  • Used in electrochemical detection.
  • Useful for organic optoelectronic devices.
  • Performs well in fluorescent probes.
  • Applicable in biosensors.
  • Utilized in photocatalytic reactions.
Catalog No. 50-004-04235 Supplier Medchemexpress LLC Supplier No. HY34515100G
Add to Cart
Add to Cart
3,4,7,8-Tetramethyl-1,10-phenanthroline (TMPhen) is an organic molecule commonly used as a ligand or catalyst. It has a wide range of applications in different fields and is utilized in research in chemistry and life sciences.
  • Applicable in organometallic chemical reactions.
  • Used in electrochemical detection.
  • Useful for organic optoelectronic devices.
  • Performs well in fluorescent probes.
  • Applicable in biosensors.
  • Utilized in photocatalytic reactions.