Learn More
Medchemexpress LLC 3,4,7,8-Tetramethyl-1,10-phenanthroline | 1660-93-1 | 99.8% | 236.31 | 25 G
SDP
Catalog No. 5000402873
Encompass_Preferred
3,4,7,8-Tetramethyl-1,10-phenanthroline (TMPhen) is an organic molecule commonly used as a ligand or catalyst. It has a wide range of applications in different fields, such as organometallic chemical reactions, electrochemical detection, and organic optoelectronic devices.
  • Used as a ligand or catalyst
  • Applications in organometallic chemical reactions
  • Applications in electrochemical detection
  • Applications in organic optoelectronic devices
  • Excellent performance in fluorescent probes
  • Excellent performance in biosensors
  • Excellent performance in photocatalytic reactions
  • Widely used in chemistry and life sciences research
Catalog No. 50-004-02873 Supplier Medchemexpress LLC Supplier No. 25GHY34515
Add to Cart
Add to Cart
3,4,7,8-Tetramethyl-1,10-phenanthroline (TMPhen) is an organic molecule commonly used as a ligand or catalyst. It has a wide range of applications in different fields, such as organometallic chemical reactions, electrochemical detection, and organic optoelectronic devices.
  • Used as a ligand or catalyst
  • Applications in organometallic chemical reactions
  • Applications in electrochemical detection
  • Applications in organic optoelectronic devices
  • Excellent performance in fluorescent probes
  • Excellent performance in biosensors
  • Excellent performance in photocatalytic reactions
  • Widely used in chemistry and life sciences research