Learn More
Medchemexpress LLC 3-[4-[(7-hydroxy-6-methyl-2,3-dihydro-1h-inden-4-yl)methyl]-3,5-dimethylpyrazol-1-yl]propanoic acid | 1092551-88-6 | 99.9% | 328.41 g/mol | C19H24N2O3 | 5 MG
SDP
Catalog No. 5000240089
Encompass_Preferred
Omzotirome is a synthetic thyromimetic research compound used to study lipid metabolism and metabolic regulation. Reported preclinical data show it increases energy expenditure and fatty acid oxidation and reduces adiposity in high-fat diet rodent models, making it a tool compound for hyperlipidaemia and metabolic research.
  • Functional analog of iodothyronines.
  • Reported to increase energy expenditure in rodent studies.
  • Enhances fatty acid oxidation in preclinical models.
  • Suitable for hyperlipidaemia and metabolic research applications.
  • High purity (≥99.9%) appropriate for biochemical assays.
  • Chemical identifiers: CAS 1092551-88-6; molecular weight 328.41 g/mol; formula C19H24N2O3.
Catalog No. 50-002-40089 Supplier Medchemexpress LLC Supplier No. HY1064255MG
Omzotirome is a synthetic thyromimetic research compound used to study lipid metabolism and metabolic regulation. Reported preclinical data show it increases energy expenditure and fatty acid oxidation and reduces adiposity in high-fat diet rodent models, making it a tool compound for hyperlipidaemia and metabolic research.
  • Functional analog of iodothyronines.
  • Reported to increase energy expenditure in rodent studies.
  • Enhances fatty acid oxidation in preclinical models.
  • Suitable for hyperlipidaemia and metabolic research applications.
  • High purity (≥99.9%) appropriate for biochemical assays.
  • Chemical identifiers: CAS 1092551-88-6; molecular weight 328.41 g/mol; formula C19H24N2O3.