Learn More
Cayman Chemical 3 4 7-TrIhydroxyIsoflavon 50mg
SDP
Catalog No. 502980806
Encompass_Preferred
A natural isoflavonoid that has antioxidant activity; produced by the metabolism of daidzein or daidzin; inhibits several signaling pathways in cells
Catalog No. 50-298-0806 Supplier Cayman Chemical Supplier No. 2093050 MG
A natural isoflavonoid that has antioxidant activity; produced by the metabolism of daidzein or daidzin; inhibits several signaling pathways in cells