Learn More
Medchemexpress LLC 3,4,8,9-tetrahydroxy-6H-benzo[c]chromen-6-one | 131086-98-1 | MFCD00954508 | 99.7% | C13H8O6 | 10MG
SDP
Catalog No. 5000260269
Encompass_Preferred
Urolithin D (3,4,8,9-tetrahydroxy-6H-benzo[c]chromen-6-one; CAS 131086-98-1) is a colonic metabolite of ellagitannins used in biochemical and cell-based research. It acts as a competitive, reversible antagonist of EphA receptors, displays antioxidant activity, and modulates lipid metabolism and fatty acid oxidation. Offered as high-purity solid and DMSO solution formats for research use.
  • High purity (>99.7%) suitable for analytical and biological assays.
  • Molecular formula C13H8O6 and molecular weight 260.20 g/mol.
  • Acts as a competitive, reversible antagonist of EphA receptors.
  • Exhibits antioxidant activity and supports studies of oxidative stress.
  • Modulates lipid metabolism and promotes fatty acid oxidation.
  • Useful for tumor, metabolic, and inflammatory disease research.
  • Available as solid and DMSO solution formats for convenient handling.
Catalog No. 50-002-60269 Supplier Medchemexpress LLC Supplier No. HY13317810MG
Urolithin D (3,4,8,9-tetrahydroxy-6H-benzo[c]chromen-6-one; CAS 131086-98-1) is a colonic metabolite of ellagitannins used in biochemical and cell-based research. It acts as a competitive, reversible antagonist of EphA receptors, displays antioxidant activity, and modulates lipid metabolism and fatty acid oxidation. Offered as high-purity solid and DMSO solution formats for research use.
  • High purity (>99.7%) suitable for analytical and biological assays.
  • Molecular formula C13H8O6 and molecular weight 260.20 g/mol.
  • Acts as a competitive, reversible antagonist of EphA receptors.
  • Exhibits antioxidant activity and supports studies of oxidative stress.
  • Modulates lipid metabolism and promotes fatty acid oxidation.
  • Useful for tumor, metabolic, and inflammatory disease research.
  • Available as solid and DMSO solution formats for convenient handling.