Learn More
AdipoGen 3-4-Aminophenyl-7-diethylam
SDP
Catalog No. 502561723
Encompass_Preferred
Chemical. CAS 36840-64-9. Formula C20H22N2O2. MW 322.4. Fluorogenic amine label used for the synthesis of fluorescent reagents. Spectral Data lambdaex = 396nm lambdaem = 480nm in DMF, Lit.
Catalog No. 50-256-1723 Supplier AdipoGen Supplier No. CDXA0718M100
Chemical. CAS 36840-64-9. Formula C20H22N2O2. MW 322.4. Fluorogenic amine label used for the synthesis of fluorescent reagents. Spectral Data lambdaex = 396nm lambdaem = 480nm in DMF, Lit.