Learn More
Chemglass Life Sciences Coupling, 3/4" Beaded Pipe, "A" 1-1/2" X "B" 7/8", 25 in-lbs MAX Torque, 3/8" Hex Nut Size
SDP
Catalog No. CG196870
Encompass_Preferred
Coupling, 3/4" Beaded Pipe, "A" 1-1/2" X "B" 7/8", 25 in-lbs MAX Torque, 3/8" Hex Nut Size. Complete coupling assembly consists of a 300 series stainless steel outer band with T-bolt, a Viton liner, and a PTFE sealing ring. Couplings allow for a maximum deflection of 4¿. PLEASE NOTE: TIGHTEN ALL COUPLINGS TO THE PROPER TORQUE SETTING.
Catalog No. CG196870 Supplier Chemglass Life Sciences Supplier No. CG196870
Coupling, 3/4" Beaded Pipe, "A" 1-1/2" X "B" 7/8", 25 in-lbs MAX Torque, 3/8" Hex Nut Size. Complete coupling assembly consists of a 300 series stainless steel outer band with T-bolt, a Viton liner, and a PTFE sealing ring. Couplings allow for a maximum deflection of 4¿. PLEASE NOTE: TIGHTEN ALL COUPLINGS TO THE PROPER TORQUE SETTING.