Learn More
Chemglass Life Sciences 3/4" Beaded Pipe to 1/2" Female NPT Aluminum Adapter
SDP
Supplier:  Chemglass Life Sciences CG196442
Encompass_Preferred
3/4" Beaded Pipe to 1/2" Female NPT Aluminum Adapter. Aluminum transition adapter from beaded pipe on the inlet/outlet of the reactor to NPT. Used to connect fluid transfer hose to the vessel. Adapters require the beaded pipe couplings for connection to the vessel. Two are required per reactor.
Catalog No. CG196442