Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-[4-(Benzyloxy)phenyl]propionic acid, 96%

Catalog No. p-7032343
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3376603 1 g
AAH3376606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3376603 Supplier Thermo Scientific Chemicals Supplier No. H3376603
Add to Cart

CAS: 50463-48-4 | C16H16O3 | 256.301 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50463-48-4
Molecular Formula C16H16O3
Molecular Weight (g/mol) 256.301
MDL Number MFCD01570264
InChI Key QTSAUVQZNADEKS-UHFFFAOYSA-N
Synonym 3-4-benzyloxyphenyl propionic acid, 3-4-benzyloxy phenyl propanoic acid, 3-4-benzyloxy phenyl propionic acid, benzyloxyphenylpropanoic acid, 3-4-benzyloxyphenyl propanoic acid, 3-4-benzyloxy-phenyl-propionic acid, benzenepropanoic acid, 4-phenylmethoxy, maybridge3_003480, 4-benzyloxy benzenepropanoic acid
PubChem CID 561372
IUPAC Name 3-(4-phenylmethoxyphenyl)propanoic acid
SMILES C1=CC=C(C=C1)COC2=CC=C(C=C2)CCC(=O)O

Specifications

Melting Point 116°C to 118°C
Quantity 1 g
Formula Weight 256.3
Percent Purity 96%
Chemical Name or Material 3-[4-(Benzyloxy)phenyl]propionic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.