Learn More
Medchemexpress LLC 3-(4-bromo-2-chlorophenyl)piperidine-2,6-dione | 2923549-60-2 | MFCD35454792 | 98.9% | 302.55 g/mol | C11H9BrClNO2 | 25mg
SDP
Catalog No. 5000270482
Encompass_Preferred
3-(4-Bromo-2-chlorophenyl)piperidine-2 6-dione is a CRBN-type E3 ubiquitin ligase ligand that can be used to prepare PROTAC
Catalog No. 50-002-70482 Supplier Medchemexpress LLC Supplier No. HYW89898925MG
3-(4-Bromo-2-chlorophenyl)piperidine-2 6-dione is a CRBN-type E3 ubiquitin ligase ligand that can be used to prepare PROTAC