Learn More
Medchemexpress LLC 3-(4-bromo-2-fluorophenyl)piperidine-2,6-dione | 2924092-66-8 | 98.6% | 286.10 g·mol⁻¹ | C11H9BrFNO2 | 50mg
SDP
Catalog No. 5000270368
Encompass_Preferred
3-(4-Bromo-2-fluorophenyl)piperidine-2 6-dione is a CRBN-type E3 ubiquitin ligase ligand that can be used to prepare PROTAC
Catalog No. 50-002-70368 Supplier Medchemexpress LLC Supplier No. HYW88897050MG
3-(4-Bromo-2-fluorophenyl)piperidine-2 6-dione is a CRBN-type E3 ubiquitin ligase ligand that can be used to prepare PROTAC