Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-[4-(Bromomethyl)phenyl]-5-methyl-1,2,4-oxadiazole 97+%, Thermo Scientific™

Catalog No. CC34808DA
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC34808DA 1 g Amber Glass Bottle
CC34808CB 250 mg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC34808DA Supplier Thermo Scientific Chemicals Supplier No. CC34808DA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 256956-42-0
Molecular Formula C10H9BrN2O
Molecular Weight (g/mol) 253.099
InChI Key BMXSEDLZMUPKSH-UHFFFAOYSA-N
Synonym 3-4-bromomethyl phenyl-5-methyl-1,2,4-oxadiazole, 1,2,4-oxadiazole, 3-4-bromomethyl phenyl-5-methyl, 1,2,4-oxadiazole,3-4-bromomethyl phenyl-5-methyl, 3-4-bromomethyl phenyl-5-methyl-1,2,4-oxadiazol
PubChem CID 7164590
IUPAC Name 3-[4-(bromomethyl)phenyl]-5-methyl-1,2,4-oxadiazole
SMILES CC1=NC(=NO1)C2=CC=C(C=C2)CBr

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 253.1
Percent Purity 97%
Chemical Name or Material 3-[4-(Bromomethyl)phenyl]-5-methyl-1, 2, 4-oxadiazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.