Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-(4-Bromophenyl)-3-(Trifluoromethyl)-3H-Diazirine

Catalog No. 90629850MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-629-850MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 90-629-850MG Supplier Sigma Aldrich Supplier No. 90629850MG
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

3-(4-Bromophenyl)-3-(trifluoromethyl)-3H-diazirine is a Suzuki-Miyaura (S-M)-compatible building block tailored for the design of photoaffinity probes (PAPs). Photoaffinity labeling (PAL) is of growing interest in chemical biology and drug discovery research for use in protein profiling; covalent inhibitors; protein-protein interactions and complexes; and target engagement, identification, or validation studies. Diazirines are desirable photoactivable groups for chemical probes; however, their installation is not always straightforward. In response, diazirine-tolerant S-M coupling conditions were developed by Ichiishi, et al, using 3-(4-bromophenyl)-3-(trifluoromethyl)-3H-diazirine and shown to be efficient and scalable with minimal perturbation of the diazirine.Reducing Limitation in Probe Design: The Development of a Diazirine-Compatible Suzuki-Miyaura Cross Coupling Reaction

Specifications

CAS 2227157-06-2
Molecular Formula C28H39N7O6S
Linear Formula C28H39N7O6S
Quantity 50 mg
Synonym (2S,4R)-1-((S)-2-(2-(2-(2-Azidoethoxy)ethoxy)acetamido)-3,3-dimethylbutanoyl)-4-hydroxy-N-(4-(4-methylthiazol-5-yl)benzyl)pyrrolidine-2-carboxamide; Crosslinker-E3 ligase ligand conjugate; Protein degrader building block for PROTAC(R) research; Template for synthesis of targeted protein degrader; VH032 conjugate
Molecular Weight (g/mol) 601.72 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.