Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-(4-Bromophenyl)-6-nitro-2H-benzo[b][1,4]oxazine

Catalog No. JRD09861G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
JRD09861G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. JRD09861G Supplier Sigma Aldrich Supplier No. JRD09861G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 5-tert-butyl-2-hydroxybenzaldehyde is a 5-substituted 2-hydroxy aromatic aldehyde. it participates in the synthesis of schiff base appended porphyrazine.5-tert-butyl-2-hydroxybenzaldehyde may be used in the synthesis of the following organic ligands:
  • 5-tert-butyl-2-hydroxybenzaldehyde thiosemicarbazone (thtb)
  • 4-tert-butyl-2-(4,5-dimethyl-1h-imidazol-2-yl)phenol
  • 2-tert-butyl-6-(4,5-diphenyl-1h-imidazol-2-yl)phenol

Specifications

Molecular Formula C14H9BrN2O3
Linear Formula C14H9BrN2O3
MDL Number MFCD28976537
Quantity 1 g
Molecular Weight (g/mol) 333.14
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.