Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-(4-Bromophenyl)isoxazole-5-carboxylic acid

Catalog No. JRD08601G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
JRD08601G 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. JRD08601G Supplier Sigma Aldrich Supplier No. JRD08601G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 4-bromobenzaldehyde diethyl acetal is also known as p-bromobenzaldehyde dimethyl acetal
  • It is employed during chemical decomposition of pladienolide d to benzylidene acetal.4-bromobenzaldehyde diethyl acetal may be used in the preparation of 4-ethynylbenzaldehyde diethylacetal

Specifications

CAS 901930-35-6
Quantity 10 mL
Molecular Formula C10H6BrNO3
Linear Formula C10H6BrNO3
MDL Number MFCD07701371
Molecular Weight (g/mol) 268.06
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.