Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(4-Bromophenyl)propiolic Acid 98.0+%, TCI America™
SDP

Catalog No. B52431G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B52431G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B52431G Supplier TCI America Supplier No. B52431G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 25294-65-9
Molecular Formula C9H5BrO2
Molecular Weight (g/mol) 225.04
MDL Number MFCD03230769
InChI Key GZFOMZGALYVQDX-UHFFFAOYSA-N
Synonym (4-Bromophenyl)propargylic Acid, 3-(4-Bromophenyl)-2-propynoic Acid
PubChem CID 825805
IUPAC Name 3-(4-bromophenyl)prop-2-ynoic acid
SMILES OC(=O)C#CC1=CC=C(Br)C=C1

Specifications

Melting Point 203°C
Quantity 1 g
Formula Weight 225.04
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-(4-Bromophenyl)propiolic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.