Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(4-Bromophenyl)pyridine 96.0+%, TCI America™
SDP

Catalog No. B46171G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B46171G 1 g
B46175G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B46171G Supplier TCI America Supplier No. B46171G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 129013-83-8
Molecular Formula C11H8BrN
Molecular Weight (g/mol) 234.10
MDL Number MFCD04116232
InChI Key FCHUOBPHXDXZBK-UHFFFAOYSA-N
PubChem CID 1515241
IUPAC Name 3-(4-bromophenyl)pyridine
SMILES BrC1=CC=C(C=C1)C1=CN=CC=C1

Specifications

Melting Point 40°C
Color White-Yellow
Quantity 1 g
Formula Weight 234.10
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-(4-Bromophenyl)pyridine
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.