Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-(4-Chlorophenyl)-7-hydroxy-4H-chromen-4-one

Catalog No. OTV0003261G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OTV0003261G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OTV0003261G Supplier Sigma Aldrich Supplier No. OTV0003261G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 2-chloro-4-(trifluoromethyl)pyrimidine is a pyrimidine derivative
  • Its density and refractive index have been determined.2-chloro-4-(trifluoromethyl)pyrimidine may be used to investigate the effect of chemical substitutions on the interfacial interactions of pyrimidines with the phospholipid-mimic immobilized-artificial-membrane (iam) chromatographic stationary phase
  • Monocyclic pyrimidine nucleic acid bases (nucleobases) were reported to behave differently from their bicyclic purine analogs

Specifications

CAS 15485-81-1
Molecular Formula C15H9ClO3
Linear Formula C15H9ClO3
MDL Number MFCD00498170
Quantity 1 g
Molecular Weight (g/mol) 272.68
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.