Learn More
Medchemexpress LLC 3-[(4-chlorophenyl)methoxy]-N-[(1S)-1-phenylethyl]thiophene-2-carboxamide | 1203680-76-5 | MFCD18086887 | >98.0% | 371.88 g/mol | C20H18ClNO2S | 10 MG
SDP
Catalog No. 5000298301
Encompass_Preferred
AS1949490 (3-[(4-chlorophenyl)methoxy]-N-[(1S)-1-phenylethyl]thiophene-2-carboxamide) is a potent, orally active, and selective inhibitor of SHIP2 phosphatase used for research into insulin signaling and metabolic disease. It increases Akt phosphorylation, glucose consumption, and glucose uptake in cellular models. Intended for research use only, it has a molecular weight of 371.88 g/mol and CAS 1203680-76-5.
  • Selective SHIP2 inhibitor with nanomolar activity for mouse and human SHIP2.
  • Promotes Akt phosphorylation and increases glucose uptake in cells.
  • Suitable for metabolic and diabetes research applications.
  • Molecular formula C20H18ClNO2S and molecular weight 371.88 g/mol.
  • Typical supplier-reported purity ≥98% (HPLC).
  • Powder stable at -20 °C; in solution stable at -80 °C (6 months) or -20 °C (1 month).
Catalog No. 50-002-98301 Supplier Medchemexpress LLC Supplier No. HY1868610MG
AS1949490 (3-[(4-chlorophenyl)methoxy]-N-[(1S)-1-phenylethyl]thiophene-2-carboxamide) is a potent, orally active, and selective inhibitor of SHIP2 phosphatase used for research into insulin signaling and metabolic disease. It increases Akt phosphorylation, glucose consumption, and glucose uptake in cellular models. Intended for research use only, it has a molecular weight of 371.88 g/mol and CAS 1203680-76-5.
  • Selective SHIP2 inhibitor with nanomolar activity for mouse and human SHIP2.
  • Promotes Akt phosphorylation and increases glucose uptake in cells.
  • Suitable for metabolic and diabetes research applications.
  • Molecular formula C20H18ClNO2S and molecular weight 371.88 g/mol.
  • Typical supplier-reported purity ≥98% (HPLC).
  • Powder stable at -20 °C; in solution stable at -80 °C (6 months) or -20 °C (1 month).