Learn More
Medchemexpress LLC 3-(4-Chloropyridin-2-yl)oxetan-3-amine hydrochloride | 2879235-73-9 | 221.08 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY200656
Encompass_Preferred
3-(4-Chloropyridin-2-yl)oxetan-3-amine hydrochloride is a chemical substance intended for laboratory research. It is classified as harmful if swallowed, and can cause skin and serious eye irritation, as well as respiratory irritation. Proper handling and storage are crucial for safe use.
  • Harmful if swallowed
  • Causes skin irritation
  • Causes serious eye irritation
  • May cause respiratory irritation
  • Store at room temperature, keep dry and cool
  • Keep container tightly sealed
  • For research use only and for experienced personnel
  • Must be handled by suitably qualified experienced scientists in appropriately equipped and authorized facilities
  • Stable under recommended storage conditions
  • Incompatible with strong acids/alkalis, strong oxidising/reducing agents
  • Under fire conditions, may decompose and emit toxic fumes
Catalog No. 50-004-02519