Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(4-Chlorosulfonylphenyl)propionic acid, 99%

Catalog No. AAH5038406
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5038406 5 g
AAH5038403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5038406 Supplier Thermo Scientific Chemicals Supplier No. H5038406
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 63545-54-0 | C9H9ClO4S | 248.68 g/mol

3-(4-Chlorosulfonylphenyl)propionic acid is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(4-Chlorosulfonylphenyl)propionic acid is used as pharmaceutical intermediate.

Solubility
It reacts with water.

Notes
Moisture sensitive. Incompatible with oxidizing agents, bases and active metals.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63545-54-0
Molecular Formula C9H9ClO4S
Molecular Weight (g/mol) 248.68
MDL Number MFCD02179510
InChI Key INJMPXHPNFJMLG-UHFFFAOYSA-N
Synonym 3-4-chlorosulfonyl phenyl propanoic acid, benzenepropanoicacid, 4-chlorosulfonyl, 3-4-chlorosulphonyl phenyl propanoic acid, 3-4-chlorosulfonylphenyl propionic acid, 3-4-chlorosulfonylphenyl propanoic acid, p-chlorosylphonyldihydrocinnamic acid, 4-chlorosulfonyl benzenepropanoic acid, 4-chlorosulphonyl dihydrocinnamic acid, p-chlorosulphonyldihydrocinnamic acid
PubChem CID 2757915
IUPAC Name 3-[4-(chlorosulfonyl)phenyl]propanoic acid
SMILES OC(=O)CCC1=CC=C(C=C1)S(Cl)(=O)=O

Specifications

Quantity 5 g
UN Number UN3261
Sensitivity Moisture sensitive
Solubility Information It reacts with water.
Formula Weight 248.69
Percent Purity 99%
Chemical Name or Material 3-(4-Chlorosulfonylphenyl)propionic acid
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.