Learn More
Hach Company 3/4 in Combination pH Sensor, Convertible Style, PPS Body, Glass Electrode, PT1000 Temperature Compensation
Catalog No. 5000207634 Shop All Hach Company Products
Encompass
The 3/4 in combination pH sensor is a convertible-style, general-purpose online probe designed for immersion or flow-through installation. It features a PPS body, a glass pH electrode with a PTFE double junction, integrated PT1000 temperature compensation, and a 4.5 m analog cable for reliable pH monitoring in water and wastewater applications.
  • Convertible mounting for immersion or flow-through use.
  • PPS body for chemical resistance and long-term durability.
  • Glass pH electrode with PTFE double junction for stable pH response.
  • Integrated PT1000 temperature compensation for accurate readings.
  • 4.5 m (15 ft) analog cable for flexible installation options.
  • FKM/FPM o-rings and compatible wetted materials for improved chemical compatibility.
  • Suitable for municipal water, industrial wastewater, and process monitoring.
Catalog No. 50-002-07634 Supplier Hach Company Supplier No. PC1R1ASTC
The 3/4 in combination pH sensor is a convertible-style, general-purpose online probe designed for immersion or flow-through installation. It features a PPS body, a glass pH electrode with a PTFE double junction, integrated PT1000 temperature compensation, and a 4.5 m analog cable for reliable pH monitoring in water and wastewater applications.
  • Convertible mounting for immersion or flow-through use.
  • PPS body for chemical resistance and long-term durability.
  • Glass pH electrode with PTFE double junction for stable pH response.
  • Integrated PT1000 temperature compensation for accurate readings.
  • 4.5 m (15 ft) analog cable for flexible installation options.
  • FKM/FPM o-rings and compatible wetted materials for improved chemical compatibility.
  • Suitable for municipal water, industrial wastewater, and process monitoring.