Learn More
Cayman Chemical 3,4-dehydro Cilostazol, Purity: ≥90%, CAS: 73963-62-9, 10 mg
SDP
Catalog No. 502911051
Encompass_Preferred
An active metabolite of cilostazol; formed from cilostazol by CYP3A4 and CYP2C19
Catalog No. 50-291-1051 Supplier Cayman Chemical Supplier No. 3507510 MG
An active metabolite of cilostazol; formed from cilostazol by CYP3A4 and CYP2C19