Learn More
Medchemexpress LLC 3,4-dehydro-L-proline | 4043-88-3 | MFCD00065964 | 99.4% | 113.11 | C5H7NO2 | 5 G
SDP
Catalog No. 5000302870
Encompass_Preferred
3,4-Dehydro-L-proline is an amino acid derivative (C5H7NO2, MW 113.11) used as a synthetic building block and ligand component in organic and peptide synthesis and in biochemical research. It is supplied as a solid with high chemical purity for research applications.
  • High purity (≈99.4%) suitable for research applications.
  • Versatile building block for organic and peptide synthesis.
  • Useful as a ligand or intermediate in medicinal chemistry research.
  • Solid form for convenient storage and handling.
  • Characterized by standard analytical data for identity confirmation.
Catalog No. 50-003-02870 Supplier Medchemexpress LLC Supplier No. HYW0158975G
3,4-Dehydro-L-proline is an amino acid derivative (C5H7NO2, MW 113.11) used as a synthetic building block and ligand component in organic and peptide synthesis and in biochemical research. It is supplied as a solid with high chemical purity for research applications.
  • High purity (≈99.4%) suitable for research applications.
  • Versatile building block for organic and peptide synthesis.
  • Useful as a ligand or intermediate in medicinal chemistry research.
  • Solid form for convenient storage and handling.
  • Characterized by standard analytical data for identity confirmation.