Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3,4-Diaminotoluene, 97%

Click to view available options
Quantity:
100g
500g
496-72-0
C7H10N2
122.171
MFCD00007728
DGRGLKZMKWPMOH-UHFFFAOYSA-N
3,4-diaminotoluene,3,4-toluenediamine,1,2-benzenediamine, 4-methyl,4-methyl-o-phenylenediamine,toluene-3,4-diamine,4-methyl-1,2-phenylenediamine,4-methyl-1,2-benzenediamine,3,4-tolylenediamine,3,4-toluylenediamine,1,2-diamino-4-methylbenzene
10332
4-methylbenzene-1,2-diamine
CC1=CC(=C(C=C1)N)N
496-72-0
C7H10N2
122.171
MFCD00007728
DGRGLKZMKWPMOH-UHFFFAOYSA-N
3,4-diaminotoluene,3,4-toluenediamine,1,2-benzenediamine, 4-methyl,4-methyl-o-phenylenediamine,toluene-3,4-diamine,4-methyl-1,2-phenylenediamine,4-methyl-1,2-benzenediamine,3,4-tolylenediamine,3,4-toluylenediamine,1,2-diamino-4-methylbenzene
10332
4-methylbenzene-1,2-diamine
CC1=CC(=C(C=C1)N)N
Quantity 500g
Boiling Point 265°C
Melting Point 86°C to 90°C
Beilstein 507965
Formula Weight 122.17
Percent Purity 97%
Chemical Name or Material 3,4-Diaminotoluene

GHS H Statement
H301-H341-H319
Toxic if swallowed.
Suspected of causing genetic defects.
Causes serious eye irritation.

GHS P Statement
P280-P281-P301+P310-P305+P351+P338-P405-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : XS9820000

TSCA : TSCA