Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Diaminotoluene, 97%

Catalog No. p-7027542
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2437822 100 g
AAB2437836 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2437822 Supplier Thermo Scientific Chemicals Supplier No. B2437822
Only null left
Add to Cart

CAS: 496-72-0 | C7H10N2 | 122.171 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 496-72-0
Molecular Formula C7H10N2
Molecular Weight (g/mol) 122.171
MDL Number MFCD00007728
InChI Key DGRGLKZMKWPMOH-UHFFFAOYSA-N
Synonym 3,4-diaminotoluene, 3,4-toluenediamine, 1,2-benzenediamine, 4-methyl, 4-methyl-o-phenylenediamine, toluene-3,4-diamine, 4-methyl-1,2-phenylenediamine, 4-methyl-1,2-benzenediamine, 3,4-tolylenediamine, 3,4-toluylenediamine, 1,2-diamino-4-methylbenzene
PubChem CID 10332
IUPAC Name 4-methylbenzene-1,2-diamine
SMILES CC1=CC(=C(C=C1)N)N

Specifications

Melting Point 86°C to 90°C
Boiling Point 265°C
Quantity 100 g
Beilstein 507965
Formula Weight 122.17
Percent Purity 97%
Chemical Name or Material 3,4-Diaminotoluene
Hazard Category H302-H319-H341
Hazard Statement GHS H Statement
H301-H341-H319
Toxic if swallowed.
Suspected of causing genetic defects.
Causes serious eye irritation.
Precautionary Statement P201-P202-P264b-P270-P280i-P281-P301+P312-P305+P351+P338-P308+P313-P330-P501c
EINECSNumber 207-826-2
RTECSNumber XS9820000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.