Learn More
Medchemexpress LLC 3,4-Dibromo-Mal-PEG2-amine TFA | 2296708-07-9 | 98.0% | C12H15Br2F3N2O6 | 100 MG
SDP
Catalog No. 5000355411
Encompass_Preferred
3,4-Dibromo-Mal-PEG2-amine TFA is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs are compounds comprising two distinct ligands linked together; one ligand targets an E3 ubiquitin ligase, while the other targets the protein of interest. This design enables PROTACs to leverage the intracellular ubiquitin-proteasome system for the selective degradation of target proteins.
  • PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Enables selective degradation of target proteins
  • Targets E3 ubiquitin ligase and protein of interest
Catalog No. 50-003-55411 Supplier Medchemexpress LLC Supplier No. HY141004A100MG
3,4-Dibromo-Mal-PEG2-amine TFA is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs are compounds comprising two distinct ligands linked together; one ligand targets an E3 ubiquitin ligase, while the other targets the protein of interest. This design enables PROTACs to leverage the intracellular ubiquitin-proteasome system for the selective degradation of target proteins.
  • PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Enables selective degradation of target proteins
  • Targets E3 ubiquitin ligase and protein of interest