Learn More
Medchemexpress LLC 3,4-Dibromo-Mal-PEG2-N-Boc | 1807537-43-4 | 95.3% | C15H22Br2N2O6 | 250 MG
SDP
Catalog No. 5000352893
Encompass_Preferred
3,4-Dibromo-Mal-PEG2-N-Boc is a PEG-based PROTAC linker used in the synthesis of PROTACs. These molecules are designed to exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • PROTACs contain two different ligands connected by a linker
  • One ligand targets an E3 ubiquitin ligase
  • The other ligand targets a specific protein
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-52893 Supplier Medchemexpress LLC Supplier No. 250MGHY141005
3,4-Dibromo-Mal-PEG2-N-Boc is a PEG-based PROTAC linker used in the synthesis of PROTACs. These molecules are designed to exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • PROTACs contain two different ligands connected by a linker
  • One ligand targets an E3 ubiquitin ligase
  • The other ligand targets a specific protein
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins