Learn More
Medchemexpress LLC 3,4-Dibromo-Mal-PEG2-N-Boc | 1807537-43-4 | 95.31% | C15H22Br2N2O6 | 50 MG
SDP
Catalog No. 5000355412
Encompass_Preferred
3,4-Dibromo-Mal-PEG2-N-Boc is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs consist of two different ligands connected by a linker; one ligand is for an E3 ubiquitin ligase and the other is for the target protein. PROTACs work by utilizing the intracellular ubiquitin-proteasome system to selectively degrade target proteins. This product appears as a solid, white to off-white in color, and is for research use only.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Utilizes intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • Appears as a solid
  • Color is white to off-white
  • For research use only
Catalog No. 50-003-55412 Supplier Medchemexpress LLC Supplier No. HY14100550MG
3,4-Dibromo-Mal-PEG2-N-Boc is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs consist of two different ligands connected by a linker; one ligand is for an E3 ubiquitin ligase and the other is for the target protein. PROTACs work by utilizing the intracellular ubiquitin-proteasome system to selectively degrade target proteins. This product appears as a solid, white to off-white in color, and is for research use only.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Utilizes intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • Appears as a solid
  • Color is white to off-white
  • For research use only