Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3,4-Dibromo-Mal-PEG8-Boc | 2055198-02-0 | 98.0% | C27H45Br2NO12 | 50 MG
SDP

Catalog No. 5000355030
Encompass_Preferred

3,4-Dibromo-Mal-PEG8-Boc is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs are compounds that facilitate the selective degradation of target proteins by leveraging the intracellular ubiquitin-proteasome system. They consist of two ligands joined by a linker, one targeting an E3 ubiquitin ligase and the other a target protein.

  • Used as a PEG-based PROTAC linker.
  • Purity: 98.0%
  • Appearance: Liquid, colorless to light yellow.
  • Solubility: Soluble in DMSO (100 mg/mL).
  • Storage (pure form): -20°C for 3 years, 4°C for 2 years.
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month.

Catalog No. 50-003-55030 Supplier Medchemexpress LLC Supplier No. HY14100750MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

3,4-Dibromo-Mal-PEG8-Boc is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs are compounds that facilitate the selective degradation of target proteins by leveraging the intracellular ubiquitin-proteasome system. They consist of two ligands joined by a linker, one targeting an E3 ubiquitin ligase and the other a target protein.

  • Used as a PEG-based PROTAC linker.
  • Purity: 98.0%
  • Appearance: Liquid, colorless to light yellow.
  • Solubility: Soluble in DMSO (100 mg/mL).
  • Storage (pure form): -20°C for 3 years, 4°C for 2 years.
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.