Learn More
Accela Chembio Inc MF: C4H2Br2S | MW: 241.93 | ?98% | BP: 221-222¿C | Density: 2.188.
Catalog No. 50353171
Encompass_Preferred
MF: C4H2Br2S | MW: 241.93 | ?98% | BP: 221-222¿C | Density: 2.188.
Catalog No. 50-353-171 Supplier Accela Chembio Inc Supplier No. SY001342100G
MF: C4H2Br2S | MW: 241.93 | ?98% | BP: 221-222¿C | Density: 2.188.