Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Dibromothiophene 98.0+%, TCI America™
SDP

Catalog No. D167625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D167625G 25 g
D16765G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D167625G Supplier TCI America Supplier No. D167625G
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3141-26-2
Molecular Formula C4H2Br2S
Molecular Weight (g/mol) 241.93
MDL Number MFCD00005465
InChI Key VGKLVWTVCUDISO-UHFFFAOYSA-N
Synonym thiophene, 3,4-dibromo, 3,4-dibromo-thiophene, pubchem5510, thiophene,4-dibromo, 3, 4-dibromothiophene, acmc-1clwy, 3,4-bis bromanyl thiophene, ksc224a6j, 3,4-dibromothiophene, bidd:gt0493
PubChem CID 18452
IUPAC Name 3,4-dibromothiophene
SMILES BrC1=CSC=C1Br

Specifications

Melting Point 5°C
Color Yellow
Boiling Point 100°C
Quantity 25 g
Formula Weight 241.93
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,4-Dibromothiophene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.